missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MEPE/OF45 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB3140
This item is not returnable.
View return policy
Description
MEPE/OF45 Monoclonal specifically detects MEPE/OF45 in Human samples. It is validated for Western Blot.
Specifications
| MEPE/OF45 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NQ76 | |
| MEPE | |
| Mouse myeloma cell line NS0-derived recombinant human MEPE/OF45 Pro18-Asp525 Accession # Q9NQ76 | |
| 100 μg | |
| Primary | |
| Detects human MEPE/OF45 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 341611 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| extracellular phosphoglycoprotein with ASARM motif (bone), matrix extracellular phosphoglycoprotein, OF45, Osteoregulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56955 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction