missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melanotransferrin/CD228 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB81751
This item is not returnable.
View return policy
Description
Melanotransferrin/CD228/MFI2 Monoclonal specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Melanotransferrin/CD228/MFI2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P08582 | |
| MFI2 | |
| HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228 Gly20-Gly711 Accession # P08582 | |
| 100 μg | |
| Primary | |
| Detects human Melanotransferrin/CD228 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 893416 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MFI2, MGC4856, MTF, MTF1, p97 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4241 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction