Learn More
Human Melanopsin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB10497-100
 View more versions of this product
Product Code. 18764407
  • £429.00 / 100µg
Description
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Melanopsin
Monoclonal
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4
NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Western Blot, Flow Cytometry, CyTOF
628737
Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Q9UHM6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
94233
Human
Purified