missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melanopsin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10497-SP
This item is not returnable.
View return policy
Description
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Melanopsin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4 | |
| NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 628737 | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9UHM6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 94233 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction