missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melanopsin Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB10497T-100UG
This item is not returnable.
View return policy
Description
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Flow Cytometry.
Specifications
| Melanopsin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4 | |
| NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 628737 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| Q9UHM6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 94233 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction