Learn More
Human Melanopsin Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB10497G-100UG
Product Code. 18778586
  • £443.00 / 100µg
Description
Melanopsin Monoclonal specifically detects Melanopsin in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Melanopsin
Monoclonal
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
melanopsin, MGC142118, MOP, OPN4, opsin 4, Opsin-4
NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6
100 μg
Primary
Human
Purified
Flow Cytometry
628737
Flow Cytometry 0.25-1 ug/106 cells
Q9UHM6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
94233
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
IgG2a