missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melan-A/MART-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8008-SP
This item is not returnable.
View return policy
Description
Melan-A/MART-1 Monoclonal specifically detects Melan-A/MART-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Melan-A/MART-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q16655 | |
| MLANA | |
| E. coli-derived recombinant human Melan-A/MART-1 Asn52-Pro118 Accession # Q16655 | |
| 25 μg | |
| Primary | |
| Detects human Melan-A/MART-1 in ELISA. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 872719 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART-1, MART1MART-1, melan-A, MelanA, melanoma antigen recognized by T-cells 1, MLANA, Protein Melan-A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2315 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction