missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MEKK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6095
This item is not returnable.
View return policy
Description
MEKK3 Monoclonal specifically detects MEKK3 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| MEKK3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| Q99759 | |
| MAP3K3 | |
| E. coli-derived recombinant human MEKK3 Ser43-Tyr275 Accession # Q99759 | |
| 100 μg | |
| Primary | |
| Detects human MEKK3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, KnockDown | |
| 576240 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.11.25, MAP/ERK kinase kinase 3, MAP3K3, MAPK/ERK kinase kinase 3, MAPKKK3, MAPKKK3MEKK 3, MEK kinase 3, MEKK3, mitogen-activated protein kinase kinase kinase 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4215 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction