missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MEGF6 (aa 444-527) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62559 (PA5-62559. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MEGF6 gene ontology annotations related to this gene include calcium ion binding.
Specifications
Specifications
| Accession Number | O75095 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1953 |
| Name | Human MEGF6 (aa 444-527) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2600001P17Rik; Egfl3; EGF-like domain-containing protein 3; EGF-like protein 3; EGF-like-domain, multiple 3; Epidermal growth factor-like protein 3; KIAA0815; MEGF6; multiple EGF like domains 6; multiple EGF-like domain protein 3; multiple EGF-like domains protein 6; multiple EGF-like-domains 6; multiple epidermal growth factor-like domains 6; multiple epidermal growth factor-like domains protein 6 |
| Common Name | MEGF6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction