missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Megalin/LRP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9578-100
This item is not returnable.
View return policy
Description
LRP2 Monoclonal specifically detects LRP2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| LRP2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| calcium sensor protein, DBS, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP2, LRP-2, megalin | |
| Mouse myeloma cell line NS0-derived recombinant human Megalin/LRP2 Pro3510-Lys3964 Accession # P98164 | |
| 100 μg | |
| 4036 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| CyTOF, Flow Cytometry, Immunohistochemistry | |
| 545606 | |
| Flow Cytometry 0.25μg/106 cells, Immunohistochemistry 5-25μg/mL, CyTOF-ready | |
| P98164 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human Megalin/LRP2 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction