Learn More
Invitrogen™ Human MED31 (aa 2-72) Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP96552
Description
Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57477 (PA5-57477. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
MED31 is the component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes.
Specifications
| Q9Y3C7 | |
| Blocking Assay, Control | |
| 51003 | |
| 100 μL | |
| 3110004H13Rik; CGI-125; fa04d01; FLJ27436; FLJ36714; hSOH1; l11Jus15; med31; Mediator complex subunit 31; mediator complex subunit soh1; Mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription subunit 31; LOW QUALITY PROTEIN: mediator of RNA polymerase II transcription subunit 31; mediator of RNA polymerase II transcription, subunit 31 homolog; RGD1309457; soh1; wu:fa04d01; zgc:92673 | |
| MED31 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human MED31 (aa 2-72) Control Fragment | |
| RUO | |
| MED31 | |
| Unconjugated | |
| Recombinant | |
| AAAVAMETDDAGNRLRFQLELEFVQCLANPNYLNFLAQRGYFKDKAFVNYLKYLLYWKDPEYAKYLKYPQC | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.