missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MED27 (aa 79-171) Control Fragment Recombinant Protein

Product Code. 30201026
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201026

Brand: Invitrogen™ RP89375

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52374 (PA5-52374. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6P2C8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9442
Name Human MED27 (aa 79-171) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500015J03Rik; 2310042P07Rik; AA682045; cofactor required for Sp1 transcriptional activation subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8, 34 kDa; CRAP34; CRSP complex subunit 8; CRSP34; CRSP8; D2Ertd434e; LOC525389 protein; MED27; MED3; mediator complex subunit 27; Mediator of RNA polymerase II transcription subunit 27; MGC11274; p37 TRAP/SMCC/PC2 subunit; RGD1564993; transcriptional coactivator CRSP34; transcriptional co-activator CRSP8 homolog; TRAP37
Common Name MED27
Gene Symbol MED27
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KPSENHPLHNSGLLSLDPVQDKTPLYSQLLQAYKWSNKLQYHAGLASGLLNQQSLKRSANQMGVSAKRRPKAQPTTLVLPPQYVDDVISRIDR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.