missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MEC (CCL28) Recombinant Protein, PeproTech®

Product Code. 17862553
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17862553

Brand: Gibco™ 300571MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MEC is a 12.3 kDa protein containing 108 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor, and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRJ3
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 56477
Molecular Weight (g/mol) 12.3 kDa
Name Human MEC (CCL28)
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CC chemokine CCL28; C-C motif chemokine 28; C-C motif chemokine ligand 28; CCK 1; CCK1; CCK1 protein; CCL 28; Ccl28; chemokine (C-C motif) ligand 28; chemokine (C-C motif) ligand 28 splice variant chi; MEC; Mucosae-associated epithelial chemokine; Protein CCK1; SCYA28; small inducible cytokine A28; small inducible cytokine subfamily A (Cys-Cys), member 28; small-inducible cytokine A28
Common Name CCL28
Gene Symbol CCL28
Biological Activity Determined by its ability to chemoattract human lymphocytes using a concentration range of 1.0-10.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SEAILPIASS CCTEVSHHIS RRLLERVNMC RIQRADGDCD LAAVILHVKR RRICVSPHNH TVKQWMKVQA AKKNGKGNVC HRKKHHGKRN SNRAHQGKHE TYGHKTPY
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.