missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MDGA2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5184
Additional Details : Weight : 0.00970kg
Description
MDGA2 Polyclonal specifically detects MDGA2 in Human samples. It is validated for Western Blot.Specifications
MDGA2 | |
Polyclonal | |
Unconjugated | |
Q7Z553 | |
MDGA2 | |
Mouse myeloma cell line NS0-derived recombinant human MDGA2 Gln21-Asp931 Accession # Q7Z553 | |
100 μg | |
Primary | |
Detects human MDGA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human MDGA1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
MAM domain containing glycosylphosphatidylinositol anchor 2, MAMDC1 | |
Sheep | |
Affinity Purified | |
RUO | |
161357 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only