missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MD-2/LY96 Recombinant Protein, PeproTech®

Product Code. 17860283
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17860283

Brand: Gibco™ 16007250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PeproTech's HEK293 cell-derived Recombinant Human MD-2/LY96 is a disulfide-linked homodimer whose monomer contains 148 amino acid residues, including a C-terminal His tag, and has a calculated molecular weight of 17.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MD-2 is a gene that encodes a protein involved in the recognition of bacterial lipopolysaccharides (LPS) by the immune system. It is located on chromosome 8 and is part of a larger family of genes that encode for similar proteins. The MD-2 protein forms a complex with Toll-like receptor 4 (TLR4) to recognize and respond to LPS, triggering an immune response. Mutations or variations in the MD-2 gene have been associated with various diseases and conditions, including sepsis and inflammatory disorders.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6Y9
For Use With (Application) ELISA, Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 23643
Molecular Weight (g/mol) 17.2 kDa
Name Human MD-2/LY96
Quantity 250 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias Esop1; ESOP-1; LPS co-receptor MD-2; LY96; ly-96; Lymphocyte antigen 96; MD2; MD-2; myeloid differentiation factor-2; myeloid differentiation protein-2; protein MD-2
Common Name MD-2
Gene Symbol LY96
Biological Activity Human MD-2/LY96 demonstrates concentration-dependent ability to immobilize rhTLR4 in a functional ELISA
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence QKQYWVCNSS DASISYTYCD KMQYPISINV NPCIELKGSK GLLHIFYIPR RDLKQLYFNL YITVNTMNLP KRKEVICRGS DDDYSFCRAL KGETVNTTIS FSFKGIKFSK GKYKCVVEAI SGSPEEMLFC LEFVILHQPN SNHHHHHH
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.