missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCM9 (aa 1073-1136) Control Fragment Recombinant Protein

Product Code. 30194025
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30194025

Brand: Invitrogen™ RP94959

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56653 (PA5-56653. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NXL9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 254394
Name Human MCM9 (aa 1073-1136) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 9030408O17Rik; C6orf61; dJ329L24.1; dJ329L24.3; DNA helicase MCM9; DNA replication licensing factor MCM9; Gm235; hMCM9; MCM9; Mcmdc1; Minichromosome maintenance 9; minichromosome maintenance 9 homologous recombination repair factor; minichromosome maintenance complex component 9; mini-chromosome maintenance deficient 9; minichromosome maintenance deficient domain containing 1; mini-chromosome maintenance deficient domain-containing protein 1; ODG4; RGD1560557
Common Name MCM9
Gene Symbol MCM9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SPPPERKNRGERGPSSPPTTTAPMRVSKRKSFQLRGSTEKLIVSKESLFTLPELGDEAFDCDWD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.