missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MCM10 (aa 286-360) Control Fragment Recombinant Protein

Product Code. 30197887
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197887

Brand: Invitrogen™ RP105551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (84%), Rat (84%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67218 (PA5-67218. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre-RC) and it may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein can interact with MCM2 and MCM6, as well as with the origin recognition protein ORC2. It is regulated by proteolysis and phosphorylation in a cell cycle-dependent manner. Studies of a similar protein in Xenopus suggest that the chromatin binding of this protein at the onset of DNA replication is after pre-RC assembly and before origin unwinding. Alternatively spliced transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7L590
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55388
Name Human MCM10 (aa 286-360) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410041F14Rik; AU018508; C330019M07Rik; C79164; CNA43; DNA43; homolog of yeast MCM10; hsMCM10; Mcm10; MCM10 minichromosome maintenance deficient 10; MGC126776; minichromosome maintenance 10 replication initiation factor; minichromosome maintenance complex component 10; minichromosome maintenance deficient 10; minichromosome maintenance deficient 10 (S. cerevisiae); PRO2249; Protein MCM10 homolog
Common Name MCM10
Gene Symbol MCM10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence REKLEEIDWVTFGVILKKVTPQSVNSGKTFSIWKLNDLRDLTQCVSLFLFGEVHKALWKTEQGTVVGILNANPMK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.