missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mcl-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB8281-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Mcl-1 Monoclonal specifically detects Mcl-1 in Human samples. It is validated for Immunohistochemistry.
Especificaciones
| Mcl-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bcl2-L-3, BCL2L3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4170 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 602901 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q07820 | |
| MCL1 | |
| E. coli-derived recombinant human Mcl-1 Met231-Arg350 Accession # Q07820 | |
| 25 μg | |
| Primary | |
| Detects human Mcl-1 in direct ELISAs. | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido