missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mCherry Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB11041G-100UG
This item is not returnable.
View return policy
Description
mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Flow Cytometry
Specifications
| mCherry | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2783C | |
| Flow Cytometry | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| Human mCherry synthetic peptide | |
| 100 μg | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction