missing translation for 'onlineSavingsMsg'
Learn More

Human MCAM/CD146 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 7 publications

£181.00 - £1795.00

Specifications

Antigen MCAM/CD146
Clone 128018
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18757139
GREENER_CHOICE
View Documents
R&D Systems
MAB932-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18798478
GREENER_CHOICE
View Documents
R&D Systems
MAB932-100
100 μg
£447.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18759537
GREENER_CHOICE
View Documents
R&D Systems
MAB932-500
500 μg
£1795.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications

Specifications

MCAM/CD146
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
Monoclonal
Mouse
Human
AAA20824
4162
Mouse myeloma cell line NS0-derived recombinant human MCAM Val24-Gly559 Accession # AAA20824
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human MCAM in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm) OCAM, or rmMAdCAM-1 is observed.
128018
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin
MCAM
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.