Learn More
R&D Systems™ human MBTPS1/SKI-1 (NP_003782.1) VersaClone cDNA
Product Code. 18770977
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity Unit Size
18770977 10 μg 10µg
1 options
Product Code. 18770977 Supplier R&D Systems™ Supplier No. RDC3632
  • ONLINE EXCLUSIVE PRICE
    £599.00
    £565.95 / 10µg
    Save £33.05
    5% Off
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Gene MBTPS1
Gene ID (Entrez) 8720
Gene Alias EC 3.4.21.112, Endopeptidase S1P, KIAA0091MGC138712, membrane-bound transcription factor peptidase, site 1, membrane-bound transcription factor protease, site 1, PCSK8, S1PMGC138711, site-1 protease, SKI1, SKI-1 membrane-bound transcription factor site-1 protease, subtilisin/kexin isozyme-1, Subtilisin/kexin-isozyme 1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.