missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MBOAT2 (aa 468-520) Control Fragment Recombinant Protein

Product Code. 30205937
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205937

Brand: Invitrogen™ RP91250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53238 (PA5-53238. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Acyltransferase which mediates the conversion of lysophosphatidylethanolamine (1-acyl-sn-glycero-3- phosphoethanolamine or LPE) into phosphatidylethanolamine (1,2- diacyl-sn-glycero-3-phosphoethanolamine or PE) (LPEAT activity). Catalyzes also the acylation of lysophosphatidic acid (LPA) into phosphatidic acid (PA) (LPAAT activity). Has also a very weak lysophosphatidylcholine acyltransferase (LPCAT activity). Prefers oleoyl-CoA as the acyl donor. Lysophospholipid acyltransferases (LPLATs) catalyze the reacylation step of the phospholipid remodeling pathway also known as the Lands cycle.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6ZWT7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 129642
Name Human MBOAT2 (aa 468-520) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1-acylglycerophosphate O-acyltransferase; 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoethanolamine O-acyltransferase; 231n7; 2810049G06Rik; AU022889; AW547221; LPAAT; LPC acyltransferase 4; LPCAT; Lpcat4; LPEAT; LPLAT 2; lyso-PA acyltransferase; lyso-PC acyltransferase 4; lyso-PE acyltransferase; lysophosphatidic acid acyltransferase; Lysophosphatidylcholine acyltransferase; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase; Lysophospholipid acyltransferase 2; MBOAT2; membrane bound O-acyltransferase domain containing 2; membrane-bound O-acyltransferase domain-containing protein 2; mLPCAT4; Moact2; OACT2; O-acyltransferase (membrane bound) domain containing 2; O-acyltransferase domain-containing protein 2
Common Name MBOAT2
Gene Symbol MBOAT2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TQRRKNTHENIQLSQSKKFDEGENSLGQNSFSTTNNVCNQNQEIASRHSSLKQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.