missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MBL2 (aa 102-240) Control Fragment Recombinant Protein

Product Code. 30211830
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211830

Brand: Invitrogen™ RP95395

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (56%), Rat (56%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110650 (PA5-110650. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MBL2 is a calcium-dependent lectin involved in innate immune defense. It binds mannose, fucose and N-acetylglucosamine on different microorganisms and activates the lectin complement pathway. It also binds to late apoptotic cells, as well as to apoptotic blebs and to necrotic cells, but not to early apoptotic cells, facilitating their uptake by macrophages. MBL2 may bind DNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11226
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4153
Name Human MBL2 (aa 102-240) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 27 kDa mannan-binding protein monomeric subunit; Ab2-001; Ab2-011; cMBl; COLEC1; collectin; Collectin1; collectin-1; c-type lectin; HSMBPC; L-MBP; mannan-binding lectin; mannan-binding lectin C; mannan-binding protein; mannose binding lectin (C); mannose binding lectin 2; mannose binding lectin 2 (protein C); mannose binding lectin, liver (A); mannose binding lectin, liver (C); mannose-binding lectin; mannose-binding lectin (protein C) 2; mannose-binding lectin (protein C) 2, soluble; mannose-binding lectin (protein C) 2, soluble (opsonic defect); mannose-binding lectin 2, soluble (opsonic defect); mannose-binding lectin C; mannose-binding lectin precursor protein; mannose-binding lectin-C; mannose-binding protein; Mannose-binding protein C; mannose-binding protein C (liver); mbl; MBL2; MBL2D; MBL-C; MBP; MBP1; MBPC; MBP-C; MBPD; MGC116832; MGC116833; pMBP-27; RA-reactive factor P28A subunit; ra-reactive factor polysaccharide-binding component p28A; RaRF p28A; RARF/P28A; serum protein; MBL; soluble mannose-binding lectin
Common Name MBL2
Gene Symbol MBL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DGDSSLAASERKALQTEMARIKKWLTFSLGKQVGNKFFLTNGEIMTFEKVKALCVKFQASVATPRNAAENGAIQNLIKEEAFLGITDEKTEGQFVDLTGNRLTYTNWNEGEPNNAGSDEDCVLLLKNGQWNDVPCSTSH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.