missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MBL Oligomer Human ELISA Kit

Product Code. 10202148
Click to view available options
Sufficient For:
96 microplate wells
Unit Size:
Each
This item is not returnable. View return policy

This product is available for purchase only in the U.S.A.

Product Code. 10202148

Brand: Invitrogen™ KIT029

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This product is available for purchase only in the U.S.A.

Sandwich ELISA Kit

The Human Mannose Binding Lectin (Hu MBL-2) ELISA quantitates Hu MBL-2 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu MBL-2. Principle of the method The Human MBL-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Mannan-binding lectin (MBL; also called mannose binding lectin or protein) is a multimeric carbohydrate-binding protein produced in the liver and secreted into the blood, where it constitutes an important element in innate immune defense against invading microorganisms. Its normally oligomerized forms are associated with specific serine proproteases (the MASPs) which are activated when MBL binds to microbial carbohydrate surfaces and in turn activate complement via the MBL or lectin pathway. The determination of MBL concentrations in serum may be useful for the elucidation of suspected immune defects and as a prognostic indicator alerting to the need for heightened therapeutic or prophylactic measures in immunosuppressed patients, including patients receiving cancer chemotherapy, and patients with cystic fibrosis, SLE or rheumatoid arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11226
Assay Range 0-40 ng/mL
Assay Sensitivity 5 pg/mL
Description Human MBL Oligomer ELISA Kit
Conjugate Biotin
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Product Type ELISA Kit
Sample Type Plasma, Serum
Specificity Western blotting of plasma or plasma fractions has not identified bands reacting with the antibody other than those attributable to MBL. The absence of cross-reaction with other plasma components is supported by the fact that readings indistinguishable fr
Sufficient For 96 microplate wells
Target Species Human
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4153
Gene Symbol COLEC1,HSMBPC,MBLD,MBP,MBP-C,MBP1,MBPD,MBL2
Intraassay CV 5.5%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer concentrate, Chromogen, Stop Solution
Label or Dye HRP
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Undetermined
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma, 10 μL; Serum, 10 μL
Storage Requirements 2°C to 8°C
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 15 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.