missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MBL Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF2307
Additional Details : Weight : 0.00970kg
Description
MBL Polyclonal specifically detects MBL in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
MBL | |
Polyclonal | |
Unconjugated | |
AAH96182 | |
MBL2 | |
Mouse myeloma cell line NS0-derived recombinant human MBL Glu21-Ile248 Accession # AAH96182 | |
100 μg | |
Primary | |
Detects human MBL in ELISAs and Western blots. In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant mouse (rm) MBL and rmMBL-2 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
COLEC1, COLEC1collectin-1, Collectin-1, HSMBPC, Mannan-binding protein, Mannose-binding lectin, mannose-binding lectin (protein C) 2, soluble, mannose-binding lectin (protein C) 2, soluble (opsonic defect), mannose-binding protein C, MBL2, MBL2D, MBLmannan-binding lectin, MBP, MBP1, MBP1mannose-binding lectin 2, soluble (opsonic defect), MBP-C, MGC116832, MGC116833 | |
Goat | |
Affinity Purified | |
RUO | |
4153 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only