missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Matriptase/ST14 Catalytic Domain Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 14 publications
Brand: R&D Systems AF3946
This item is not returnable.
View return policy
Description
Matriptase/ST14 Polyclonal specifically detects Matriptase/ST14 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Matriptase/ST14 | |
| Polyclonal | |
| Unconjugated | |
| Q9Y5Y6 | |
| ST14 | |
| E. coli-derived recombinant human Matriptase/ST14 Catalytic Domain Val615-Val855 Accession # Q9Y5Y6 | |
| 100 μg | |
| Primary | |
| Detects human Matriptase/ST14 Catalytic Domain in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| EC 3.4.21, Epithin, HAI, Matriptase, Membrane-type serine protease 1, MTSP1, MT-SP1EC 3.4.21.109, prostamin, PRSS14, Serine protease 14, Serine protease TADG-15, SNC19, SNC19MTSP1, ST14, suppression of tumorigenicity 14 (colon carcinoma), suppression of tumorigenicity 14 (colon carcinoma, matriptase, epithin), suppressor of tumorigenicity 14 protein, TADG15, TADG-15, TMPRSS14, tumor associated differentially expressed gene 15 protein, Tumor-associated differentially-expressed gene 15 protein | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| 6768 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction