missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Matrilin-3 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
Description
Human Matrilin-3 quantitates human Matrilin-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Matrilin-3.
Matrilins (MATNs) are a family of non-collagenous extra-cellular matrix (ECM) proteins consisting of four known members that have been proposed to play key roles in modulating cellular phenotypes during chondrogenesis of mesenchymal stem cells (MSCs). MATN1 and MATN3 are expressed specifically in cartilage and are among the most up-regulated ECM proteins during chondrogenesis. MATN3 is composed of a single N-terminal von Willebrand Factor A (vWFA) domain followed by four epidermal growth factor (EGF) repeats and a coiled-coil domain whereas MATN1 is composed of two vWFA domains separated by one EGF-like domain. MATN1 or MATN3 may play a role in modulating chondrogenesis during the chondrocyte differentiation process. Mutations of this gene have been associated with variety of inherited chondrodysplasias. Recent studies show that aberrant expression and processing of MATN3 are hallmarks of conventional cartilaginous neoplasms.
Specifications
Specifications
| Accession Number | AV009181; B14.5 b; CI-B14.5 b; DIPOA; EDM5; HOA; Matn3; matrilin 3; matrilin-3; NADHDH2; NDUFC2; OADIP; OS2 |
| Assay Range | 20 to 5,000 pg/mL |
| Assay Sensitivity | 20 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 4148 |
| Gene Symbol | MATN3 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction