missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Matrilin-3 Antibody, R&D Systems™
Goat Polyclonal Antibody
£418.32
Specifications
Antigen | MATN3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Applications | Western Blot, Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
MATN3 Polyclonal antibody specifically detects MATN3 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
MATN3 | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
4148 | |
Mouse myeloma cell line NS0-derived recombinant human Matrilin-3 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Matrilin-3 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Matrilin-3 is observed and less than 1% cross-reactivity with recombinant human (rh) Matrilin-2 and rhMatrilin-4 is observed. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
DIPOA, EDM5, HOA, matrilin 3, matrilin-3, Matrilin3, NDUFC2, OADIP, OS2 | |
NDUFC2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only