missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MARCH4 (aa 312-396) Control Fragment Recombinant Protein

Product Code. 30197437
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30197437

Brand: Invitrogen™ RP90983

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53149 (PA5-53149. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

RNF174 (MARCH4) is a multi-pass membrane protein mainly found in the membrane of Golgi apparatus. It is a complex enzyme with a RING-CH-type zinc finger. It acts as a ubiquitin-protein ligase mainly involved in third step of ubiquitin conjugation. The RINGv-type zinc finger domain is required for E3 ligase activity in which the E3 ubiquitin-protein ligase mediates ubiquitination of MHC-I and CD4, and promote their subsequent endocytosis and sorting to lysosomes via multivesicular bodies. It is mainly expressed in brain and placenta.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9P2E8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 57574
Name Human MARCH4 (aa 312-396) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC056494; E3 ubiquitin-protein ligase MARCH4; Kiaa1399; MARCH4; MARCHF4; MARCH-IV; membrane associated ring finger 4; membrane associated ring-CH-type finger 4; membrane-associated ring finger (C3HC4) 4; membrane-associated ring finger (C3HC4) 4, E3 ubiquitin protein ligase; membrane-associated RING finger protein 4; Membrane-associated RING-CH protein IV; mKIAA1399; RING finger protein 174; RING-type E3 ubiquitin transferase MARCH4; RNF174
Common Name MARCH4
Gene Symbol MARCH4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence WKVLNYDKTKDLEDQKAGGRTNPRTSSSTQANIPSSEEETAGTPAPEQGPAQAAGHPSGPLSHHHCAYTILHILSHLRPHEQRSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.