missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MARCH3 (aa 204-253) Control Fragment Recombinant Protein

Product Code. 30180933
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30180933

Brand: Invitrogen™ RP99910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60351 (PA5-60351. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the membrane-associated RING-CH (MARCH) family. The encoded protein is an E3 ubiquitin-protein ligase that may be involved in regulation of the endosomal transport pathway. [provided by RefSeq, Mar 2013].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86UD3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 115123
Name Human MARCH3 (aa 204-253) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6330411I15Rik; A530081L18Rik; AI843639; E3 ubiquitin-protein ligase MARCH3; MARCH3; MARCH-III; membrane associated ring finger 3; membrane associated ring-CH-type finger 3; membrane-associated ring finger (C3HC4) 3; membrane-associated ring finger (C3HC4) 3, E3 ubiquitin protein ligase; membrane-associated RING finger protein 3; Membrane-associated RING-CH protein III; RING finger protein 173; RING-type E3 ubiquitin transferase MARCH3; RNF173
Common Name MARCH3
Gene Symbol MARCH3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FRYHCRLYNEWRRTNQRVILLIPKSVNVPSNQPSLLGLHSVKRNSKETVV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.