missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAPKAP1 (aa 155-303) Control Fragment Recombinant Protein

Product Code. 30193872
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193872

Brand: Invitrogen™ RP89609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83062 (PA5-83062. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Sin1 was initially identified as the human homolog of S. pombe SIN1. Recent evidence has shown that it identical to Mip1, a protein that interacts with MEKK2, a member of the mitogen-activated protein kinase (MAPK) intracellular signaling network. MAPKAP1 is thought to prevent MEKK2 activation and dimerization by forming a complex with the inactive and non-phosphorylated MEKK2, thereby blocking the JNK1/2, ERK1/2, p38 and ERK5 MAPKs. MAPKAP1 has also been shown to play a role in the TOR signaling process, a pathway that is involved in controlling cell growth and proliferation in response to environmental cues such as nutrients, growth factors and hormones. Experiments showed that MAPKAP1 helped to maintain the TOR/rictor assembly but not the TOR/RAPTOR complex, which allowed specific phosphorylation of Akt, a kinase that is believed to couple the growth factor-PI3K signaling pathway to the nutrient-regulated TOR signaling pathway. Multiple alternatively spliced isoforms of MAPKAP1 have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BPZ7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79109
Name Human MAPKAP1 (aa 155-303) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI591529; D230039K05Rik; im:7143088; JC310; MAPK associated protein 1; mapkap1; MEKK2-interacting protein 1; MGC2745; Mip1; MIP1SIN1SIN1b; mitogen activated protein kinase associated protein 1; mitogen-activated protein kinase 2-associated protein 1; mitogen-activated protein kinase associated protein 1; mSIN1; ras inhibitor MGC2745; RP11-269P11.1; SAPK interacting protein; SAPK-interacting protein 1; si:ch73-242f23.1; SIN1; SIN1b; SIN1g; stress-activated map kinase interacting protein 1; stress-activated map kinase-interacting protein 1; stress-activated protein kinase-interacting 1; target of rapamycin complex 2 subunit MAPKAP1; TORC2 subunit MAPKAP1
Common Name MAPKAP1
Gene Symbol MAPKAP1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NPFNEYSKFDGKGHVGTTATKKIDVYLPLHSSQDRLLPMTVVTMASARVQDLIGLICWQYTSEGREPKLNDNVSAYCLHIAEDDGEVDTDFPPLDSNEPIHKFGFSTLALVEKYSSPGLTSKESLFVRINAAHGFSLIQVDNTKVTMKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.