missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP1A (aa 1913-2024) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66845 (PA5-66845. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Structural protein involved in the filamentous cross-bridging between microtubules and other skeletal elements.
Specifications
Specifications
| Accession Number | P78559 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 4130 |
| Name | Human MAP1A (aa 1913-2024) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 6330416M19Rik; AI853608; MAP1 light chain LC2; MAP1A; MAP-1 A; MAP1A heavy chain; MAP1L; microtubule associated protein 1 A; microtubule-associated protein 1 A; microtubule-associated protein 1 A; modifier of tubby hearing 1; moth1; Mtap1; Mtap-1; Mtap1a; Proliferation-related protein p80 |
| Common Name | MAP1A |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido