missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MANF Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF3748
This item is not returnable.
View return policy
Description
MANF Polyclonal specifically detects MANF in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| MANF | |
| Polyclonal | |
| Unconjugated | |
| P55145 | |
| MANF | |
| Mouse myeloma cell line NS0-derived recombinant human MANF Leu22-Leu179 Accession # P55145 | |
| 100 μg | |
| Primary | |
| Detects human MANF in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Knockout Validated | |
| Arginine-Rich Protein, arginine-rich, mutated in early stage tumors, ARMET, ARP, mesencephalic astrocyte-derived neurotrophic factor, Protein ARMET | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7873 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction