missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAN1A1 Partial ORF (NP_005898.2, 110 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004121-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16074365

  • £283.00 / 10µg
Estimated Shipment: 08-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the removal of 3 distinct mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases. [provided by RefSeq]

Sequence: PGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKI
Specifications

Specifications

NP_005898.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKI
RUO
MAN1A1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4121
MAN1A1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUMM3/HUMM9/MAN9
MAN1A1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers