missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Mammaglobin B (aa 23-93) Control Fragment Recombinant Protein

Product Code. 30193568
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193568

Brand: Invitrogen™ RP95119

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (35%), Rat (35%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56981 (PA5-56981. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Secretoglobins (also called lipophilins or mammaglobins) are small secreted proteins of ndocrine-responsive organs and mucosal epithelia that form multimeric complexes and correlate with the development of various human cancers. Lipophilin A and Lipophilin B are orthologs of prostatein estramustine- binding protein), the major secretory glycoprotein of the rat ventral prostate gland. Lipophilin A, also designated LIPA, LPHA and secretoglobin, family 1D, member 1 (SCGB1D1), is a component of a heterodimeric molecule present in human tears. Lipophilin B, also designated LIPB, LPHB and secretoglobin, family 1D, member 2 (SCGB1D2), mRNA can be overexpressed in breast tumors and shows a high degree of correlation with the mRNA expression profile of mammaglobin. Histological detection in breast tissue of Mammaglobin A, also designated MGB1 and secretoglobin, family 2A, member 2 (SCGB2A2) and Mammaglobin B, also designated MGB2, Lipophilin C, LPHC, UGB3 and SCGB2A2, is a reliable diagnostic marker for breast tumors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75556
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4246
Name Human Mammaglobin B (aa 23-93) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C3.1; Lacryglobin; LIPHC; lipophilin C; lipophilin-C; LPHC; LPNC; mammaglobin 2; mammaglobin B; mammaglobin-2; Mammaglobin-B; MGB2; prostatein; Prostatein peptide C3; Prostatic steroid-binding protein C3; Psbp1; Psbpc3; SCGB2A1; Scgb2a2; Secretoglobin family 2 A member 1; Secretoglobin family 2 A member 2; secretoglobin, family 2 A, member 1; UGB3
Common Name Mammaglobin B
Gene Symbol SCGB2A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KLLEDMVEKTINSDISIPEYKELLQEFIDSDAAAEAMGKFKQCFLNQSHRTLKNFGLMMHTVYDSIWCNMK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.