missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MALT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4850-SP
This item is not returnable.
View return policy
Description
MALT1 Monoclonal specifically detects MALT1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| MALT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q9UDY8 | |
| MALT1 | |
| Recombinant human MALT1 Asp701-Thr808 Accession # Q9UDY8 | |
| 25 μg | |
| Primary | |
| Detects human MALT1 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| MALT1-C | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| caspase-like protein, DKFZp434L132, EC 3.4.22.-, MALT associated translocation, MALT lymphoma-associated translocation, MLT1, MLTMALT-lymphoma associated translocation, mucosa associated lymphoid tissue lymphoma translocation gene 1, mucosa-associated lymphoid tissue lymphoma translocation protein 1, paracaspase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10892 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction