missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MAGEA10 (aa 44-150) Control Fragment Recombinant Protein

Product Code. 30203194
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30203194

Brand: Invitrogen™ RP92198

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (40%), Rat (40%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51897 (PA5-51897. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

MAGEA10 is a member of the MAGEA gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 80% sequence identity between each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are expressed at a high level in a number of tumors of various histologic types, and are silent in normal tissues with the exception of testis and placenta. The MAGEA genes are clustered on chromosome Xq28. They may be implicated in some hereditary disorders, such as dyskeratosis congenita.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43363
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 4109
Name Human MAGEA10 (aa 44-150) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cancer/testis antigen 1.10; cancer/testis antigen family 1, member 10; CT1.10; MAGE family member A10; MAGE10; MAGE-10 antigen; MAGEA10; melanoma antigen family A, 10; melanoma antigen family A10; melanoma-associated antigen 10; RGD1563693
Common Name MAGEA10
Gene Symbol MAGEA10
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TSSSFPSSFPSSSSSSSSSCYPLIPSTPEEVSADDETPNPPQSAQIACSSPSVVASLPLDQSDEGSSSQKEESPSTLQVLPDSESLPRSEIDEKVTDLVQFLLFKYQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.