missing translation for 'onlineSavingsMsg'
Learn More

Human MAG/Siglec-4a Antibody, R&D Systems™

Product Code. 30689911
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689911 25 μg 25µg
30689960 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30689911 Supplier R&D Systems Supplier No. MAB11688SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MAG/Siglec-4a Monoclonal antibody specifically detects MAG/Siglec-4a in Human samples. It is validated for Western Blot,Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAG/Siglec-4a
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1099521
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Alias GMAsialic acid binding Ig-like lectin 4A, myelin associated glycoprotein, myelin-associated glycoprotein, sialic acid-binding immunoglobulin-like lectin 4A, Siglec4a, SIGLEC-4A, S-MAG
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human MAG/Siglec-4a, Gly20-Pro516
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4099
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.