missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human M1AP (aa 32-128) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66244 (PA5-66244. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in gametogenesis.
Specifications
Specifications
| Accession Number | Q8TC57 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 130951 |
| Name | Human M1AP (aa 32-128) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C2orf65; D6Mm5e; M1AP; Meiosis 1 arrest protein; meiosis 1 arresting protein; meiosis 1 associated protein; meiosis 1-arresting protein; Meiosis 1-associated protein; RGD1563842; Spata37; spermatogenesis associated 37; spermatogenesis-associated protein 37 |
| Common Name | M1AP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction