missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LYVE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB11463-100
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal antibody specifically detects LYVE-1 in Human samples. It is validated for Flow Cytometry
Specifications
| LYVE-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1, UNQ230/PRO263, XLKD1 | |
| Mouse myeloma cell line NS0-derived recombinant human LYVE-1, Ser24-Thr238, Accession # Q9Y5Y7 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry | |
| 1072614 | |
| Flow Cytometry 0.25 μg/10^6 cells | |
| Q9Y5Y7 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10894 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction