missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LYVE-1 Alexa Fluor™ 750-conjugated Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11463S-100UG
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal antibody specifically detects LYVE-1 in Human samples. It is validated for Flow Cytometry
Specifications
| LYVE-1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Q9Y5Y7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10894 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1072614 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1, UNQ230/PRO263, XLKD1 | |
| Mouse myeloma cell line NS0-derived recombinant human LYVE-1, Ser24-Thr238, Accession # Q9Y5Y7 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction