missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysyl Oxidase Homolog 3/LOXL3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6069-100
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 3/LOXL3 Monoclonal specifically detects Lysyl Oxidase Homolog 3/LOXL3 in Human samples. It is validated for ELISA.
Specifications
| Lysyl Oxidase Homolog 3/LOXL3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 1.4.3, EC 1.4.3.-, EC 1.4.3.13, LOL3, LOXL, LOXL3, lysyl oxidase homolog 3, lysyl oxidase-like 3, Lysyl oxidase-like protein 3 | |
| Chinese Hamster Ovary cell line CHO-derived human Lysyl Oxidase Homolog 3/LOXL3 protein, Met1-Ile753, Accession # P58215 | |
| 100 μg | |
| 84695 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 1013328 | |
| ELISA | |
| P58215 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Lysyl Oxidase Homolog 3/LOXL3 protein in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction