missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysyl Oxidase Homolog 2/LOXL2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2639
This item is not returnable.
View return policy
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| EC 1.4.3, EC 1.4.3.-, LOL2, LOR2, LOXL2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4017 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunoprecipitation | |
| 262418 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LOXL2 | |
| Mouse myeloma cell line NS0-derived recombinant human Lysyl Oxidase Homolog 2/LOXL2 Gln26-Gln744 (predicted) | |
| 100 μg | |
| Primary | |
| Detects human Lysyl Oxidase Homolog 2/LOXL2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction