missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysine (K)-specific Demethylase 5B/KDM5B Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6477
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Polyclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
| Polyclonal | |
| Unconjugated | |
| Q9UGL1 | |
| KDM5B | |
| E. coli-derived recombinant human KDM5B Leu819-Arg918 Accession # Q9UGL1 | |
| 100 μg | |
| Primary | |
| Detects human KDM5B in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CT31, CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, Jarid1B, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), KDM5B, lysine (K)-specific demethylase 5B, Lysine (K)specific Demethylase 5B, PLU-1, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10765 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction