missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysine (K)-specific Demethylase 4C/KDM4C Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6430
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Polyclonal specifically detects Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C | |
| Polyclonal | |
| Unconjugated | |
| Q9H3R0 | |
| KDM4C | |
| E. coli-derived recombinant human KDM4C Lys381-Lys588 Accession # Q9H3R0 | |
| 100 μg | |
| Primary | |
| Detects human KDM4C in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human KDM4A is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| EC 1.14.11, EC 1.14.11.-, GASC1, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KDM4C, KIAA0780FLJ25949, lysine (K)-specific demethylase 4C, Lysine (K)specific Demethylase 4C, lysine-specific demethylase 4C | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23081 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction