missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LYPLAL1 Full-length ORF (NP_620149.1, 1 a.a. - 237 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16197743
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16197743 25 μg 25µg
16187743 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16197743 Supplier Abnova™ Supplier No. H00127018P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

Sequence: MAAASGSVLQRCIVSPAGRHSASLIFLHGSGDSGQGLRMWIKQVLNQDLTFQHIKIIYPTAPPRSYTPMKGGISNVWFDRFKITNDCPEHLESIDVMCQVLTDLIDEEVKSGIKKNRILIGGFSMGGCMAMHLAYRNHQDVAGVFALSSFLNKASAVYQALQKSNGVLPELFQCHGTADELVLHSWAEETNSMLKSLGVTTKFHSFPNVYHELSKTELDILKLWILTKLPGEMEKQK

Tekniske data

Accession Number NP_620149.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 127018
Molecular Weight (g/mol) 52.7kDa
Name LYPLAL1 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen MAAASGSVLQRCIVSPAGRHSASLIFLHGSGDSGQGLRMWIKQVLNQDLTFQHIKIIYPTAPPRSYTPMKGGISNVWFDRFKITNDCPEHLESIDVMCQVLTDLIDEEVKSGIKKNRILIGGFSMGGCMAMHLAYRNHQDVAGVFALSSFLNKASAVYQALQKSNGVLPELFQCHGTADELVLHSWAEETNSMLKSLGVTTKFHSFPNVYHELSKTELDILKLWILTKLPGEMEKQK
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ99730/KIAA1238
Common Name LYPLAL1
Gene Symbol LYPLAL1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.