missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LYPD1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6855-SP
This item is not returnable.
View return policy
Description
LYPD1 Polyclonal specifically detects LYPD1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| LYPD1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ41033, LY6/PLAUR domain containing 1, ly6/PLAUR domain-containing protein 1, LYPDC1, MGC29643, PHTS, Putative HeLa tumor suppressor | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 116372 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| Q8N2G4 | |
| LYPD1 | |
| E. coli-derived recombinant human LYPD1 Cys54-Ser116 Accession # Q8N2G4 | |
| 25 μg | |
| Primary | |
| Detects human LYPD1 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human LYPD3 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction