missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lymphotoxin-alpha/TNF-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB211-SP
This item is not returnable.
View return policy
Description
Lymphotoxin-alpha/TNF-beta Monoclonal specifically detects Lymphotoxin-alpha/TNF-beta in Human samples. It is validated for Neutralization, Immunocytochemistry.
Specifications
| Lymphotoxin-alpha/TNF-beta | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.1-0.2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P01374 | |
| LTA | |
| E. coli-derived recombinant human Lymphotoxin-alpha/TNF-beta Leu35-Leu205 Accession # P01374 | |
| 25 μg | |
| Primary | |
| Detects human Lymphotoxin-alpha/TNF-beta in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) TNF-alpha is observed. Neutralizes the biological activity of recombinant human TNF-beta . Will not neutralize the biological activity of recombinant human TNF-alpha or recombinant mouse TNF-alpha . | |
| Human | |
| Purified |
| Neutralization, Immunocytochemistry | |
| 5802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| LT, LTA, LT-alpha, Lymphotoxin alpha, lymphotoxin alpha (TNF superfamily, member 1), Lymphotoxinalpha, tnfb, TNF-beta, TNFBlymphotoxin-alpha, TNFSF1, TNFSF1B, TNFSF1TNF-beta, tumor necrosis factor beta, Tumor necrosis factor ligand superfamily member 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4049 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction