missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LYAR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6748-SP
This item is not returnable.
View return policy
Description
LYAR Monoclonal specifically detects LYAR in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| LYAR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9NX58 | |
| LYAR | |
| E. coli-derived recombinant human LYAR Lys288-Lys379 Accession # Q9NX58 | |
| 25 μg | |
| Primary | |
| Detects human LYAR in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 735838 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cell growth-regulating nucleolar protein, FLJ20425, Ly1 reactive homolog (mouse), PNAS-5, ZLYAR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55646 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu