missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LY6G5C (aa 6-64) Control Fragment Recombinant Protein

Product Code. 30202595
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202595

Brand: Invitrogen™ RP96588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (29%), Rat (29%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58495 (PA5-58495. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

LY6G5C belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q5SRR4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80741
Name Human LY6G5C (aa 6-64) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C6orf20; Canine epididymal gene product 8; CE8; CE8 protein, variant 2; epididymal secretory protein 8; G5C; Ly6g5c; LY6G5CA; LY6G5CB; Lymphocyte antigen 6 complex locus protein G5c; lymphocyte antigen 6 complex, locus G5C; lymphocyte antigen 6 family member G5C; lymphocyte antigen-6 G5C; NG33; re8
Common Name LY6G5C
Gene Symbol Ly6g5c
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ADLPSCWGAGPCYTGHKVGALRRDTVICCCRHGDYSTPCLFTPGKPSRNPSPWKRTLWT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.